Learn More
RUBIS High Precision Tweezers - SwissTweeze
Click to view available options
Length (Metric):
110 mm
115 mm
120 mm
Tip Style:
Needle Pointed, Ultra Fine
Pointed, Fine
Pointed, Ultra Fine
Round, Flat, Fine
Design:
Angled
Angled, Bent
Curved
Straight
Unit Size:
Each
Description
- High Precision Tweezers
- 100% Made in Switzerland
- Anti-acid/Antimagnetic Stainless Steel
- Perfect alignment
- Very effective
- Reliable
- Excellent chemical resistance
- Swiss Tweeze series
- Perforated handle for enhanced grip
Specifications
Specifications
| Tip Style | Pointed, Ultra Fine |
| Design | Curved |
| Length (English) | 4.52 in. |
| Material | Anti-acid, Anti-magnetic Stainless Steel |
| Length (Metric) | 115 mm |
| Product Type | High Precision Tweezers |
| Quantity (No. per Case) | Each |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.