Learn More
Sampling Systems™ Stainless Steel Pointed Scissors
Product Code. p-7146967 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
150 mm
200 mm
Length (Metric) Blade:
45 mm
100 mm
Unit Size:
Each
Product Code. 15879641
Brand: Sampling Systems™ A364150
These stainless steel scissors are made from hygienic 420 grade stainless steel with a double point design.
  • Polished finish
  • Two sizes available
  • 420 grade stainless steel
Specifications
Length (Metric) 150 mm
Length (Metric) Blade 45 mm
Material Stainless Steel (AISI 420)
Tip Style Double Point