Learn More
Caframo™ Pitched Blade Propellers, Blade Only
Click to view available options
Diameter (Metric):
2.5 cm
Includes:
8 mm Bore, Set Screw
Unit Size:
Each
Description
- Electro-polished stainless steel 316L
- Chemically inert and corrosion resistant 316L stainless steel
- For low shear, high top-to-bottom pumping capacity
- Scale up advantage if propeller used in production
Order Info
Shafts sold separately
Specifications
Specifications
| Diameter (Metric) | 2.5 cm |
| For Use With (Equipment) | Caframo™ Overhead Stirrer |
| Includes | 8 mm Bore, Set Screw |
| Type | Pitched Blade Propeller |
| Material | Stainless Steel (316L) |