Learn More
Bochem™ Stainless Steel Pharmacists Spatula
Click to view available options
Length (Metric):
190 mm
230 mm
310 mm
Length (Metric) Blade:
100 mm
130 mm
200 mm
Unit Size:
Each
Description
- Wooden handle
- Long blade
Specifications
Specifications
| Quantity | 1 Ea. |
| Product Type | Pharmacists Spatula |
| Length (Metric) Blade | 100 mm |
| Width (Metric) | 18 mm |
| Width (Metric) Blade | 18 mm |
| Blade Material | Stainless Steel |
| Handle Material | Wood |
| Material | Stainless Steel |
| Handle Type | Wood |
| Length (Metric) | 190 mm |