Learn More
TFA Dostmann™ Analog Glass Thermometer
Description
- Glass frame
- Scale with temperature markings
- High accuracy due to a 2-point calibration
- Marking of ideal temperatures
- Manufactured in Germany
Specifications
Specifications
| Calibration Points | 2-point calibration |
| Dimensions (L x W x H) | 17 x 17 x 270 mm |
| For Use With (Application) | Ideal for making butter, curds and cheese |
| Material | Glass |
| Model | 14.1006 |
| Weight (Metric) | 22 g |
| Description | Thermometer |