Learn More
Sartorius™ Stainless Steel OIML Class E2 Calibration Weight Without DAkkS Calibration Certificate
Product Code. 10309730 Shop All Sartorius Products
Change view
Click to view available options
Mass:
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mass unitSize
10309730 100 g Each
10309540 10 g Each
10627681 20 g Each
10399530 2 g Each
10379820 2 kg Each
10238180 1 kg Each
10688251 200 g Each
10591911 1 g Each
10166270 500 g Each
10298080 50 g Each
10106180 5 g Each
10084140 5 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 10309730 Supplier Sartorius™ Supplier No. YCW512800
Only null left
Add to basket
Add to basket
Highly polished steel calibration weights from 1g to 2kg
To OIML Class E2, F1 and F2 specification.
TRUSTED_SUSTAINABILITY
Specifications
Case Material Plastic
Mass 100 g