Learn More
Amsta Rostfritt Stainless Steel Measure Jug Without Lid Graduated
Product Code. p-200113436
Click to view available options
Capacity (Metric):
0.5 L
1 L
1.5 L
2 L
Unit Size:
Each
Product Code. 30284898
Brand: Amsta Rostfritt 507160,50
Only null left
Add to basket
Add to basket
Highest quality stainless steel measure jug. Manufactured in Sweden.
  • Stainless steel 304 /1.4301.
  • High quality straight graded measuring jugs.
  • Silver soldered handles to avoid gaps and places for dirt to hide and thus meet the highest standards of hygiene.
Specifications
Capacity (Metric) 0.5 L
Material Stainless Steel
Type Jug
Graduated Yes
Style Handled