missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, Aermet 100, K92580, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, Aermet 100, K92580, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Oxygen/Nitrogen/Niobium/Phosphorus/Tungsten/Aluminium/Vanadium/Titanium/Copper/Manganese/Silicon/Carbon/Molybdenum/Chromium/Nickel/Cobalt/Boron/Hafnium/Hydrogen/Tantalum/Antimony/Arsenic/Bismuth/Cadmium/Calcium/Lead/Magnesium/Selenium/Tin/Zinc/Zirconium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Oxygen 0.0009 %/Nitrogen 0.001 %/Niobium 0.003 %/Phosphorus 0.004 %/Tungsten 0.006 %/Aluminium 0.007 %/Vanadium 0.007 %/Titanium 0.008 %/Copper 0.01 %/Manganese 0.013 %/Silicon 0.039 %/Carbon 0.222 %/Molybdenum 1.19 %/Chromium 2.98 %/Nickel 11.2 %/Cobalt 13.4 %/Boron (<10) %/Hafnium (<10) %/Hydrogen (<10) %/Tantalum (<400) %/Antimony (<50) %/Arsenic (<50) %/Bismuth (<50) %/Cadmium (<50) %/Calcium (<50) %/Lead (<50) %/Magnesium (<50) %/Selenium (<50) %/Tin (<50) %/Zinc (<50) %/Zirconium (<60) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction