Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Needle, Custom length, Point Style 2, 3, or 4
Click to view available options
Needle Gauge:
10 G
12 G
13 G
14 G
15 G
16 G
22s G
23 G
24 G
26 G
26s G
27 G
28 G
33 G
Packungsgröße:
Pack of 6
Beschreibung
- Available in various gauges
- Autoclavable
- Metal hubs manufactured from nickel plated brass
- Custom length
- Compatible with PTFE Luer Lock (TLL) syringes
- Design controlled by a series of universal standards which guarantees compatibility between manufacturers
- Provided in packs of 6
Spezifikation
Spezifikation
| Autoclavable | Autoclavable |
| Material | Nickel-plated Brass (Hub), Stainless Steel |
| Needle Gauge | 16 G |
| Diameter (Metric) Inner | 1.19 mm |
| Diameter (Metric) Outer | 1.65 mm |
| Product Type | Luer Lock Custom Needle |
| Quantity | 6/Pk. |
| Length (Metric) Needle | 9.52 to 304 mm |
| Needle Point Style | 2, 3, or 4 |
| For Use With (Equipment) | TLL Syringes and LT or TLL Connectors |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur