Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Can be used with removable (RN)-style syringes that are 250μL and larger.
163.00€ - 366.00€
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Stainless Steel |
Product Type | Removable Custom Needle |
For Use With (Equipment) | Removable Syringes (250μL and larger) |
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
10146383
|
Hamilton™
7732-01 |
22 G |
366.00€
Pack of 6 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
11542525
|
Hamilton™
7806-02 |
22s G |
166.00€
Pack of 6 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
10268047
|
Hamilton™
7806-05 |
23 G |
163.00€
Pack of 6 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
10402205
|
Hamilton™
7806-01 |
22 G |
163.00€
Pack of 6 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
12316309
|
Hamilton™
7732-08 |
23s G |
366.00€
Pack of 6 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
11550042
|
Hamilton™
7806-03 |
26 G |
166.00€
Pack of 6 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
10051173
|
Hamilton™
7806-04 |
26s G |
163.00€
Pack of 6 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
12326459
|
Hamilton™
7806-09 |
25s G |
163.00€
Pack of 6 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
Description
- Available in various gauges
- Provided in packs of 6
- Custom point style and lengths
- Autoclavable
- For large hubs
Specifications
Autoclavable | |
Stainless Steel | |
Removable Custom Needle | |
Removable Syringes (250μL and larger) |