Learn More
De Buyer™ Stainless Steel Ladle with Ergonomic Handle
Product Code. 11947435
Change view
Click to view available options
Volume (Metric):
0.13 L
0.67 L
1 L
2.06 L
Length (Metric):
140 mm
340 mm
430 mm
635 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Length (Metric) Unit Size
11947435 1 L 430 mm Each
11967435 2.06 L 635 mm Each
11973674 0.13 L 340 mm Each
11903684 0.67 L 140 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11947435 Supplier De Buyer™ Supplier No. 3021.16
  • ONLINE EXCLUSIVE PRICE
    48.08€
    37.10€ / Pack of 1
    Save 10.98€
    22% Off
View all promotions
Estimated Shipment: 10-12-2026
Grip the perfectly angled ergonomic handle shape on the De Buyer™ Stainless Steel Ladle with Ergonomic Handle that lets you insert your thumb.
  • Wide and rigid 2.5mm handle
  • Fixed hook at end of handle
  • Litres capacity engraved on handle
  • Dishwasher safe
Specifications
Material Stainless Steel
Capacity (Metric) 1 L
Height (Metric) 53 mm
Length (Metric) 430 mm
Volume (Metric) 1 L