Learn More
Bochem™ Stainless Steel Laboratory Set
Product Code. 10024814
Click to view available options
Product Type:
Laboratory Set
Unit Size:
Each
Product Code. 10024814
Brand: Bochem™ 33999
Only null left
Add to basket
Add to basket
Includes four instruments in a practical Laboratory-Set. BOCHEM™ Stainless Steel Laboratory Set is ideal for the first works in the laboratory.
  • Made of 18/10 stainless steel
  • Includes one each of the following:
    • Dressing scissor (4100)
    • Micro spatula, double (3017)
    • Forceps, sharp (1012)
    • Micro scoop, half rounded (3303)
Specifications
For Use With (Application) Ideal for the first works in the laboratory
Product Type Laboratory Set