Learn More
Bochem™ Stainless Steel Laboratory Pot
Product Code. 10258320
Click to view available options
Capacity (Metric):
2.5 L
3.5 L
5 L
6.5 L
8.5 L
15.5 L
Unit Size:
Each
Product Code. 10258320
Brand: Bochem™ 8363
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Note:
Lid is not included; sold separately.
Specifications
Diameter (Metric) Inner 220 mm
Height (Metric) 176H
Capacity (Metric) 6.5 L
Material Stainless Steel (18/10)
Product Type Laboratory Pot