Learn More
Bochem™ Stainless Steel Laboratory Bowl
Product Code. 10779155
Change view
Click to view available options
Capacity (Metric):
5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10779155 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10779155 Supplier Bochem™ Supplier No. 8607
  • ONLINE EXCLUSIVE PRICE
    44.46€
    33.75€ / Pack of 1
    Save 10.71€
    24% Off
Estimated Shipment: 30-11-2026
Bochem™ high form, conical-shaped stainless steel bowls are ideal for use as mixing bowls or for general lab use.
  • High Form
  • 18/10 Stainless Steel
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
  • Wide range of uses.
Specifications
Material Stainless Steel (18/10)
Capacity (Metric) 5 L
Height (Metric) 130 mm
Diameter (Metric) 265 mm
Shape High, Round