Learn More
Rogo Sampaic™ Stainless Steel Kocher Forceps
Specifically designed to catch the bleeder that are deep within tissue. Rogo Sampaic™ Stainless Steel Kocher Forceps are good for tough structures like palms, soles or scalp.
Brand: Rogo Sampaic™ FML004
Additional Details : Weight : 0.36000kg
Description
- 160mm in length
- Supplied in packs of 10
Specifications
Kocher Forceps | |
For dissection | |
160 mm |
Stainless Steel | |
10/Pk. |