Learn More
Socorex™ Stainless Steel Injection Needle
3.10€ - 19.65€
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Stainless Steel |
Product Type | Injection Needle |
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
15203168
|
Socorex™
371.1210UN |
1.2 mm |
3.10€
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
12699285
|
Socorex™
371.1020 |
1 mm |
19.65€
Pack of 12 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Specifications
Autoclavable | |
Stainless Steel | |
Injection Needle |