Learn More
RETSCH Stainless Steel Grinding Balls
Product Code. 10539711 Shop All Retsch Products
Click to view available options
For Use With (Equipment):
MM 200 and MM 301
Planetary Ball Mill PM 400
Reaction Vials
Diameter (Metric):
3 mm
4 mm
5 mm
15 mm
40 mm
Unit Size:
Each
Pack of 200
Product Code. 10539711
Brand: RETSCH 05.368.0034
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed for use with Retsch™ mixer mills. Retsch™ Stainless Steel Grinding Balls are ideal for disruption of biological cells as well as for DNA/RNA extraction.
  • For use with MM 200 and MM 400 Mixer Mills
Specifications
Product Type Grinding Balls
Diameter (Metric) 5 mm
For Use With (Equipment) MM 200 and MM 301
Material Stainless Steel