Learn More
Aalborg™ Stainless Steel Flowmeter Glass Tube
Product Code. 15350614
Click to view available options
Housing Material:
Stainless Steel
Unit Size:
Each
Product Code. 15350614
Brand: Aalborg™ PMR1018121
Only null left
Add to basket
Add to basket
Designed for low flowrates, the model P flowmeter is a precision instrument embodying the inherent simplicity, versatility and economy of the classical variable area meter. Aalborg™ Stainless Steel Flowmeter Glass Tube is particularly suitable for metering carrier gases in chromatography.
  • Rib-guided or fluted metering tubes facilitate stable and accurate readings
  • Magnifier lens in front shield to enhance reading resolution
  • Interchangeability of Flow Tubes and floats
  • Ease of installation and exchange of flow tubes
  • Glass flow tubes are prevented from turning during the tightening phase of the assembly procedure
  • Chemical compatibility
  • Simple means of panel mounting
Specifications
Type Flowmeter
Accuracy ±5%
Pressure Rating 13.8 bar
Housing Material Stainless Steel
Width (Metric) 150 mm
Seal Material Viton-A™
Temperature (Metric) 121°C