Learn More
Grant Instruments™ Stainless Steel Flask Clamp
Click to view available options
For Use With (Equipment):
1000mL Flask
100mL Flask
250mL Flask
25mL Flask
500mL Flask
50mL Flask
Unit Size:
Each
Description
- Spring clamp provides security while allowing easy flask insertion/removal
- For use with OLS and LSB Model Aqua Pro Shaking Water Baths
Specifications
Specifications
| For Use With (Equipment) | 250mL Flask |
| Type | Flask Clamp |
| Material | Stainless Steel |
Product Content Correction
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto