Learn More
Grant Instruments™ Stainless Steel Flask Clamp
Product Code. 15365852
Click to view available options
For Use With (Equipment):
1000mL Flask
100mL Flask
250mL Flask
25mL Flask
500mL Flask
50mL Flask
Unit Size:
Each
Product Code. 15365852
Brand: Grant Instruments™ SC100
Holds flasks in place when used with Grant Aqua Pro Shaking Baths
  • Spring clamp provides security while allowing easy flask insertion/removal
  • For use with OLS and LSB Model Aqua Pro Shaking Water Baths
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 100mL Flask
Type Flask Clamp
Material Stainless Steel