Learn More
Usbeck Laborgeraete™ Stainless Steel Fine Point Forceps
Specifications
Specifications
| Tip Style | Fine |
| No. per Pack | 1 Pack |
| Material | Stainless Steel |
| Length (Metric) | 160 mm |
| Dimensions (L x W) | 160 L mm |
| Type | Forceps |