Learn More
Scotsman™ Stainless Steel EF103 Ice Flaker
Product Code. 12883216
Change view
Click to view available options
Capacity (Metric):
30 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12883216 30 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12883216 Supplier Scotsman™ Supplier No. EF103
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ice flakes are extruded at a temperature just below zero degrees and contain 20% residual water content. The flake ice is stored inside an insulated, plastic lined storage bin with front opening for easy access. Electrical supply 230V/50Hz.
  • Up to 108kg (238 lbs.) of flake ice production per day
  • Up to 30kg (66 lbs.) built-in ice storage
  • Electronically controlled operation, with self-diagnostic functions with external alarm lights providing operational information for early alert and fast diagnosis of operating issues
  • External components constructed from corrosion-resistant stainless steel crafted for optimal aesthetic appeal through superior fit and finish
  • Ergonomically convenient, slanted & disappearing storage bin door
  • Front access for easier maintenance
  • Front-in, front-out airflow allows for built-in installations
  • CFC-free R-404A refrigerant
  • 120mm (4 12/16 in.) legs included, extensions available as option
  • Two years manufacturer warranty
  • Built -in PWD (pumped waste discharge) system for remote water drain (via easy-fit installation) for discharge of waste up to 1.5m above base or up to 20m from unit Scotsman flaked ice is ideal for a variety of applications within laboratory and industrial environments.
Specifications
Capacity (Metric) 30 kg