Learn More
Bochem™ Thermo Scientific™ Stainless Steel Dressing Scissors
Product Code. 10088360
Click to view available options
Length (Metric):
130 mm
140 mm
155 mm
Tip Style:
Round/Round
Sharp/Round
Sharp/Sharp
Unit Size:
Each
Product Code. 10088360
Brand: Bochem™ 4103
Manufactured from stainless steel. Bochem™ Thermo Scientific™ Stainless Steel Dressing Scissors comes in three types.
  • Three types: round-round, sharp-round, and sharp-sharp
  • Stainless steel
Specifications
Product Type Dressing Scissors
Length (Metric) 130 mm
Material Stainless Steel
Tip Style Sharp/Sharp