Learn More
JP Selecta™ Stainless Steel Dissecting Straight Forceps
Product Code. 12022774
Change view
Click to view available options
Length (Metric):
140 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12022774 140 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12022774 Supplier JP Selecta™ Supplier No. 1000580
  • ONLINE EXCLUSIVE PRICE
    7.00€
    2.88€ / Pack of 1
    Save 4.12€
    58% Off
In Stock
Add to basket
Edge
Add to basket
JP Selecta™ Stainless Steel Dissecting Straight Forceps are suitable for laboratory purposes.
  • Made of AISI 304 stainless steel
Specifications
Product Type Dissecting Straight Forceps
Material Stainless Steel
Length (Metric) 140 mm