Learn More
Fisherbrand™ Stainless Steel Seamless, Cylindrical Scoops
Product Code. 11598092 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
1500 mL
Unit Size:
Each
Product Code. 11598092
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
These crevice-free, stainless steel, seamless scoops with a bright finish are particularly suitable for pharmaceutical applications.
  • Crevice free, stainless steel, seamless, 316 spec, bright finish
  • 316 spec stainless steel seamless cylindrical scoop
  • Particularly suitable for pharmaceutical applications 
  • All surfaces are non-reacting and offer zero absorption suiting GMP requirement
  • Produced to a durable design
  • All surfaces highly polished making cleaning easier
  • Can be sterilized by autoclaving
Specifications
Capacity (Metric) 1500 mL
Product Type Cylindrical Scoop
Length (Metric) Handle 125 mm
Material Stainless Steel
Length (Metric) 360 mm