Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class E2
Shop All Fisherbrand ProductsDescription
- Class E2 stainless steel calibration weight
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
| Classification | E2 |
| Weight (Metric) | 1 g to 5 kg |
| Mass | 1 g to 5 kg |
| Shape | Cylindrical |