Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class E2 with Certificate
Product Code. 15575119 Shop All Fisherbrand Products
Change view
Click to view available options
Weight (Metric):
1 g to 1 kg
1 g to 5 kg
1 to 100 g
200 g
Unit Size:
1 unit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) unitSize
15575119 200 g 1 unit
15575129 1 g to 5 kg 1 unit
15595119 1 g to 1 kg 1 unit
15525109 1 to 100 g 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15575119
Add to basket
Add to basket
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
  • Delivered with a calibration certificate guaranteeing traceability
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Classification E2
Weight (Metric) 200 g
Mass 200 g