Learn More
Linker Industrietechnik™ Stainless Steel Bottom for Sieving Machine
Click to view available options
Diameter (Metric) Outer:
100 mm
Unit Size:
Each
Description
- Silent screening
- Quick release
- Circular and vertical movement
Specifications
Specifications
| Quantity | 1Each |
| Type | Collecting Pan |
| Material | Stainless Steel |
| For Use With (Application) | For 100 dia. x 35mm Internal Height seives |
| Diameter (Metric) Outer | 100 mm |