Learn More
Amsta Rostfritt Stainless Steel Chem-Tech Scoop - Acid Proof
Product Code. 30284886
Change view
Click to view available options
Capacity (Metric):
4 L
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
30284886 4 L Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30284886 Supplier Amsta Rostfritt Supplier No. 517554
Only null left
Add to basket
Add to basket
Highest quality stainless steel Chem-Tech scoop - acid proof. Made of 1 mm stainless steel (EN1.4404). Intended for antibiotics. vaccines. chemicals and liquids. Manufactured in Sweden.
  • Highest quality in stainless steel.
  • Acid proof 316L/1.4404.
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 4 L
Product Type Scoop
Material Stainless Steel (316L)