Learn More
Amsta Rostfritt Stainless Steel Bucket With Lid
Click to view available options
Capacity (Metric):
10 L
12 L
15 L
Unit Size:
Each
Pack of 10
Description
- Stainless steel 304 /1.4301
Specifications
Specifications
| Product Type | Bucket with Lid |
| Capacity (Metric) | 10 L |
| Graduations | Yes |
| Material | Stainless Steel |