Learn More
Memmert™ Stainless Steel Bottom Drip Tray for UF110, 15 mm
Product Code. 17528895 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UF110 Oven
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17528895 UF110 Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17528895 Supplier Memmert™ Supplier No. UF110B04359
Non-perforated bottom drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) UF110 Oven
Type Bottom Drip Tray
Material Stainless Steel
Height (Metric) Rim: 15 mm