Learn More
Memmert Stainless steel bottom drip tray, 15mm (for reinforced chamber option), for ICH750
Brand:  Memmert ICH750B34055
Stainless steel bottom drip tray, 15mm (for reinforced chamber option), for ICH750
Product Code. 16376687
  • ONLINE EXCLUSIVE PRICE
    406.00€
    369.00€ / Each
    Save 37.00€
    9% Off
Only null left
Estimated Shipment: 17-03-2026