Learn More
Stainless Steel Blunt End Spring Forceps
Product Code. 12750916
Click to view available options
Tip Style:
Blunt
Unit Size:
Each
Product Code. 12750916
Brand: S Murray SAMCO E003/05
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Blunt End Spring Forceps are fully guaranteed in normal use and subject to stringent quality inspection procedures.
Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Blunt End Spring Forceps are fully guaranteed in normal use and subject to stringent quality inspection procedures.
  • Standard instruments are finished to a bright mirror polish
  • Stainless Steel
  • Length: 200mm
Specifications
Product Type Spring Forceps
Material Stainless Steel
For Use With (Application) For dissecting
Tip Style Blunt
Length (Metric) 200 mm