Learn More
Edmund Bühler GmbH™ Stainless Steel Beaker
Click to view available options
Capacity (Metric):
15 mL
50 mL
Unit Size:
Each
Description
- Sample vessel
- Stainless steel construction
Specifications
Specifications
| Capacity (Metric) | 15 mL |
| Material | Stainless Steel |
| Type | Beaker |