Learn More
Scientific Industries Stainless Steel Beads
Click to view available options
Diameter (Metric) Probe:
2.4 mm
3.2 mm
5.0 mm
Unit Size:
Pack of 100
Description
- Can be cleaned and reused
Specifications
Specifications
| Product Type | Beads |
| Diameter (English) Probe | 0.1259 in. |
| Diameter (Metric) Probe | 3.2 mm |
| Material | Stainless Steel |
| For Use With (Application) | Bead Beating |
| For Use With (Equipment) | Bead Genie |
| Model | SI-BS03 |
| Packaging Quantity | 100 Pk. |