Learn More
JP Selecta™ Wire Baskets
Product Code. 12092754
Change view
Click to view available options
Diameter (Metric) Outside:
270 mm
360 mm
440 mm
Height (Metric) Exterior:
200 mm
220 mm
280 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Height (Metric) Exterior Unit Size
12092754 360 mm 280 mm Each
13403669 270 mm 200 mm Each
12022804 440 mm 220 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12092754 Supplier JP Selecta™ Supplier No. 1000496
  • ONLINE EXCLUSIVE PRICE
    436.00€
    257.65€ / Pack of 1
    Save 178.35€
    40% Off
Estimated Shipment: 10-12-2026
Manufactured from AISI 304 stainless steel.
  • With electro-bright finish
  • Designed to fit Selecta autoclaves
  • Available in a range of sizes
Specifications
Autoclavable Autoclavable
Diameter (Metric) Outside 360 mm
Height (Metric) Exterior 280 mm
Product Type Basket
Features 4 Divider compartments basket
Material Stainless Steel
Quantity 1 Basket