Learn More
Zwiebel™ Stainless Steel Antimagnetic Weights Handling Clamp
Product Code. p-8001329
Click to view available options
Length (Metric):
10 cm
15 cm
Unit Size:
Each
Product Code. 11758214
Brand: Zwiebel™ PBI2812E
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these clamps for the handling of Zwiebel™ weights.
  • Stainless steel handling clamp
  • The pieces in contact with the weight are protected rubber
  • Available in a variety of sizes
Specifications
Material Anti-magnetic Stainless Steel
Length (Metric) 10 cm
Product Type Handling Clamp
For Use With (Application) For handling weights of 1mg to 50g
Quantity 1Each
Weight (Metric) 13g