missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 410, S41000, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Stainless Steel, AISI 410, S41000, 100g
Specifications
Specifications
| Chemical Name or Material | Niobium/Aluminium/Tungsten/Cobalt/Phosphorus/Copper/Nitrogen/Vanadium/Carbon/Molybdenum/Nickel/Silicon/Manganese/Chromium/Hydrogen/Lead/Tin/Bismuth/Boron/Cadmium/Magnesium/Selenium/Zinc/Zirconium/Calcium/Antimony/Tantalum/Titanium/Arsenic/Oxygen/Iron |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Niobium 0.0021 %/Aluminium 0.003 %/Tungsten 0.008 %/Cobalt 0.012 %/Phosphorus 0.017 %/Copper 0.046 %/Nitrogen 0.046 %/Vanadium 0.065 %/Carbon 0.132 %/Molybdenum 0.146 %/Nickel 0.28 %/Silicon 0.29 %/Manganese 0.5 %/Chromium 12.2 %/Hydrogen (<0.0004) %/Lead (<0.0010) %/Tin (<0.0040) %/Bismuth (<0.0050) %/Boron (<0.0050) %/Cadmium (<0.0050) %/Magnesium (<0.0050) %/Selenium (<0.0050) %/Zinc (<0.0050) %/Zirconium (<0.0050) %/Calcium (<0.0070) %/Antimony (<0.0080) %/Tantalum (<0.0130) %/Titanium (0.002) %/Arsenic (0.0032) %/Oxygen (0.009) %/Iron (86.6) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction