Learn More
Sampling systems Stainless Spoon 304 SS - 300mm long - 2ml
Brand:  Sampling systems A504-3
Stainless Spoon 304 SS - 300mm long - 2ml
Product Code. 17782229
  • ONLINE EXCLUSIVE PRICE
    45.56€
    32.53€ / Each
    Save 13.03€
    28% Off