Learn More
Brady™ StainerBondz™ Slide Labels
Product Code. 13158276
Change view
Click to view available options
Labels per Row:
4
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Labels per Row unitSize
13158276 4 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13158276 Supplier Brady™ Supplier No. 804787
Only null left
Add to basket
Add to basket
Made from one of the most solvent-resistant materials in the lab industry
  • Made of B-481 material and coupled with R6400 series ribbon
  • White matte polyester
  • Remains resistant when exposed to xylene and other chemicals commonly used in the slide staining process
  • Bar codes remain readable and labels stay adhered to the slide
  • No print removal, no label buckling, and no discoloration
  • Brady IP™ enabled
Applications:
Slide Label Identification
Specifications
Certifications/Compliance RoHS 2002/95/EC, MCV 2005/618/EC
Description Thermal Transfer Printable Labels
Adhesive Type Acrylic, Permanent
Color White
Display Consignment
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) General identification, Slide label ID
For Use With (Equipment) BBP™ 11, Brady 1244, Brady 1344 Printer
Labels per Row 4
Quantity 2500/Roll
Shape Square
Temperature Range (Metric) -80°C to +130°C
Thickness (Metric) 0.084 mm
Dimensions (L x W) 0.9 x 0.9 in. (22.86 x 22.86 mm)
Length (English) 0.9 in.
Width (English) 0.9 in.
Width (Metric) 2.28 cm
Height (English) 0.9 in.
Height (Metric) 2.28 cm
Diameter (English) 1 in.
Diameter (Metric) 2.54 cm
Alternate Part Number BPT-207-481