Learn More
Brady™ StainerBondz™ Microscope Slide Matte Polyester Labels
Artikelnummer. 17871719 Alle ansehen Brady Produkte
Ansicht ändern
Click to view available options
Dimensions (L x W):
22.86 mm (W) x 19.05 mm (H)
22.86 mm (W) x 22.86 mm (H)
22.86 mm (W) x 9.53 mm (H)
26.67 mm (W) x 6.35 mm (H)
Packungsgröße:
Pack of 280
Pack of 320
Pack of 360
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Dimensions (L x W) Packungsgröße
17871719 22.86 mm (W) x 9.53 mm (H) Pack of 320
17821789 26.67 mm (W) x 6.35 mm (H) Pack of 360
17811799 22.86 mm (W) x 19.05 mm (H) Pack of 320
17851759 22.86 mm (W) x 22.86 mm (H) Pack of 280
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17871719 Lieferant Brady™ Lieferanten-Nr. 170885
  • ONLINE EXCLUSIVE PRICE
    57.65€
    54.91€ / Pack of 320
    Save 2.74€
    4% Off
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Easily create chemical-resistant microscope slide labels with the StainerBondz™ polyester label material (B-481). Print almost anywhere with a Brady™ handheld label printer.
  • Available in presized shapes for commonly used slides and tissue cassettes in which superior chemical resistance is required
  • Withstands the rigors of typical deparaffinization including ethanol and xylene, as well as hematoxylin and eosin slide staining processes
  • Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces in extreme conditions
Spezifikation
Autoclavable Autoclavable
Certifications/Compliance 2011/65/EU, 2015/863
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Satin
Format Die-cut
Material Polyester
Printable Sides Top
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Healthcare and Hospital labelling;General identification;Laboratory labelling
For Use With (Equipment) BMP51;BMP53;M511;M510
Min. Operating Temperature (Metric) -80°C
Model M5-195-481
No. per Pack 320 Label(s) / Box
Quantity 320
Size 22.86 mm (W) x 9.53 mm (H)
Temperature Range (English) -112°F to 266°F
Temperature Range (Metric) -80°C to 130°C
Thickness (English) 0.0036 in
Thickness (Metric) 0.0914 mm
Dimensions (L x W) 22.86 mm (W) x 9.53 mm (H)
Length (English) 0.375 in.
Width (English) 0.9 in
Width (Metric) 22.86 mm
Width (English) Printable Area 22.86 mm
Height (English) 0.375 in
Height (English) Printable Area 0.5 in.
Height (Metric) 9.53 mm
Alternate Part Number M5-195-481
Mehr anzeigen Weniger anzeigen