Learn More
Scotch-Brite™ Stain Eraser 4000R
Product Code. 17108780
Click to view available options
Color:
White
Unit Size:
Pack of 30
Product Code. 17108780
Brand: Scotch-Brite™ 7100157864
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
An effective cleaning tool that can be used to remove stains or dirt easily, and effortlessly from a wide variety of surfaces
  • Easily erases stains and dirt without leaving scratches
  • High cleaning performance without the use of chemicals
  • Can be used on most common surfaces
  • Soft and flexible
  • There is no need to wash off after use
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Tool
Color White
Dimensions (L x W) 110 x 71 mm
For Use With (Application) Stain/Dirt Removal
Length (Metric) 110 mm
Quantity 30 Pk.
Width (Metric) 71 mm