Learn More
Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials
Product Code. 10346162
Change view
Click to view available options
Turbidity:
<0.1, 20, 200, 1000, 4000 NTU
<0.1, 20, 200, 1000, 4000, 7500 NTU
For Use With (Equipment):
2100AN/AN IS
2100N/N IS
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Turbidity For Use With (Equipment) Unit Size
10346162 <0.1, 20, 200, 1000, 4000, 7500 NTU 2100AN/AN IS 1 set
10070654 <0.1, 20, 200, 1000, 4000 NTU 2100N/N IS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10346162 Supplier Hach™ Supplier No. 2659505
  • ONLINE EXCLUSIVE PRICE
    809.40€
    703.34€ / 1 sets
    Save 106.06€
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-12-2026
Developed for use in any turbidimeter. Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials are true Formazin dilutions in individual sealed vials of <0.1, 20, 200, 1000, 4000 NTU and 7500 NTU (2662105 only) levels.
  • Precise, Pre-mixed Formazin Solutions: Provides accurate turbidimeter calibrations
  • Based on Formazin Primary Standards: Contains the same light scattering polymer and performs equivalently
  • Enhanced Stability Saves Time and Labor: Repeatable results without preparing standards in the lab
  • Confidence in Results: Based on decades of experience in the drinking water industry
  • Any Turbidity Meter: Can be used with 2100N/2100AN or 2100N/2100AN IS Turbidimeter
TRUSTED_SUSTAINABILITY
Specifications
Type Stablcal Turbidity Standards Calibration Kit
For Use With (Equipment) 2100AN/AN IS
Includes Premixed, sealed vials
For Use With (Application) Calibration of turbidimeters
Turbidity <0.1, 20, 200, 1000, 4000, 7500 NTU
compliance-icons
Product Identifier
  • StablCal sealed vials calibration kits
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P363-Wash contaminated clothing before reuse.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: methenamine; hexamethylenetetramine, methenamine; hexamethylenetetramine , methenamine; hexamethylen