Learn More
IKA™ ST-20-M Gamma Tubes
Shop All IKA Products
Click to view available options
For Use With (Equipment):
ST-20-M Dispersing Tool
Unit Size:
Pack of 20
Specifications
Specifications
| Product Type | ST-20-M Tube |
| Volume Range (Metric) | 2 to 15 mL |
| For Use With (Application) | Mixing, stirring, extractions, preparation of soil sample suspensions, solubility testing for medication |
| For Use With (Equipment) | ST-20-M Dispersing Tool |
| Packaging Quantity | 20 Pk. |