Learn More
IKA™ ST-20-M Gamma Tubes
Product Code. 11427296 Shop All IKA Products
Click to view available options
For Use With (Equipment):
ST-20-M Dispersing Tool
Unit Size:
Pack of 20
Product Code. 11427296
Brand: IKA™ 0003700500
Only null left
Specifications
Product Type ST-20-M Tube
Volume Range (Metric) 2 to 15 mL
For Use With (Application) Mixing, stirring, extractions, preparation of soil sample suspensions, solubility testing for medication
For Use With (Equipment) ST-20-M Dispersing Tool
Packaging Quantity 20 Pk.