Learn More
Novus Biologicals™ SSH3 293T Cell Transient Overexpression Lysate
Product Code. 18314109 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18314109 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18314109 Supplier Novus Biologicals™ Supplier No. H00054961T02
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Overexpression Lysate
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Target SSH3
Synonym EC 3.1.3.16, EC 3.1.3.48, FLJ10928, FLJ20515, FLJ42240, hSSH-3L, protein phosphatase Slingshot homolog 3, slingshot 3, slingshot homolog 3 (Drosophila), SSH3, SSH3L, SSH-3L, SSH-like protein 3
Gene Symbol SSH3