Learn More
Abnova™ SSB Recombinant Protein
Brand:  Abnova™ H00006741-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16161002
  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH01289
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
70.62
8
Glutathione Sepharose 4 Fast Flow
25 ug
MAENGDNEKMAALEAKICHQIEYYFGDFNLPRDKFLKEQIKLDEGWVPLEIMIKFNRLNRLTTDFNVIVEALSKSKAELMEISEDKTKIRRSPSKPLPEVTDEYKNDVKNRSVYIKGFPTDATLDDIKEWLEDKGQVLNIQMRRTLHKAFKGSIFVVFDSIESAKKFVETPGQKYKETDLLILFKDDYFAKKNEERKQNKVEAKLRAKQEQEAKQKLEEDAEMKSLEEKIGCLLKFSGDLDDQTCREDLHILFSNHGEIKWIDFVRGAKEGIILFKEKAKEALGKAKDANNGNLQLRNKEVTWEVLEGEVEKEALKKIIEDQQESLNKWKSKGRRFKGKGKGNKAAQPGSGKGKVQFQGKKTKFASDDEHDEHDENGATGPVKRAREETDKEEPASKQQKTENGAGDQ
LARP3/La
SSB
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
6741
SSB (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SSB
Human
Recombinant
Solution