missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SRT 1720

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SRT 1720 is selective SIRT 1 activator (EC50 = 0.16 μM). SRT 1720 inhibits growth of bladder cancer organoids and xenografts (IC50 = 0.35 μM) and induces autophagy. In vitro, SRT 1720 increases mitochondrial biogenesis and recovery after oxidation injury and reduces palmitate-induced insulin resistance. Also improves survival and healthspan of obese mice.
Specifications
Specifications
| Chemical Name or Material | N-[2-[3-(1-Piperazinylmethyl)imidazo[2,1-b]thiazol-6-yl]phenyl]-2-quinoxalinecarboxamide |
| CAS | 925434-55-5 |
| Quantity | 50 mg |
| Target | Sirtuin 1/SIRT1 |
| Molecular Formula | C25H23N7OS |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction