Learn More
TERUMO SR STRAIGHT FEP Intravenous Catheters SURFLO™
Click to view available options
Needle Gauge:
14 G
16 G
18 G
20 G
Length (English) Needle:
1.25 in.
2 in.
2.5 in.
Unit Size:
Pack of 50
Description
- The Surflo I.V. Catheters are made out of medical grade EFTE
- The materials of the Surflo™ I.V. Catheters are carefully selected to ensure a stable form during punctures under difficult conditions
- Short PIVC with an ETFE catheter tube and stainless steel internal needle
- Precisely tapered and siliconised catheter for smooth advancement of the catheter
- Catheter with X-ray contrast strips for higher patient safety when placement goes wrong
- The flashback chamber confirms needle tip entry into the vessel
- The large range in catheters and related obturators allows most appropriate choice of product for a wide range of applications
Specifications
Specifications
| Material | ETFE, Stainless Steel (Needle) |
| Color | Gray |
| Needle Gauge | 16 G |
| Latex Free | Yes |
| Length (English) Needle | 2 in. |
| Length (Metric) | 51 mm |
| Product Type | IV Catheter |
| Disposable | Single-use |
| For Use With (Equipment) | IV Lines |
| Connection Type | Leur Slip |
| Show More |