missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 16832

Description
SR 16832 is a dual site PPARγ inhibitor. Acts at orthosteric and allosteric sites in the ligand binding domain. Inhibits binding of endogenous ligands and transcriptional activity of PPARγ, more effectively than the orthosteric covalent antagonists GW 9662 (Cat. No. 1508) and T 0070907 (Cat. No. 2301).
Specifications
Specifications
| CAS | 2088135-12-8 |
| Synonym | 2-Chloro-N-(6-methoxy-4-quinolinyl)-5-nitrobenzamide |
| Quantity | 25 mg |
| Molecular Formula | C17H12ClN3O4 |
| Formula Weight | 357.75 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?