missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SR 11237
GREENER_CHOICE

Product Code. 18035172
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18035172

Brand: Tocris Bioscience™ 3411/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pan RXR agonist

Pan retinoid X receptor (RXR) agonist that is devoid of any RAR activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146670-40-8
Molecular Formula C24H28O4

Specifications

Quantity 50 mg
Formula Weight 380.48
Purity >98%
Molecular Weight (g/mol) 380.484
Chemical Name or Material SR 11237
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.